Assay Details
Gene Symbol: MYB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121666.3 | NP_197163.1 | 1 - 2 | 257 | 150 | |
GenBank mRNA | AF175990.1 | - | 1 - 2 | 131 | 150 | |
AY519625.1 | - | 1 - 2 | 131 | 150 | ||
BT002762.1 | - | 1 - 2 | 131 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
