Assay Details
Gene Symbol: MYB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121666.3 | NP_197163.1 | 2 - 3 | 385 | 154 | |
GenBank mRNA | AF175990.1 | - | 2 - 3 | 259 | 154 | |
AY519625.1 | - | 2 - 3 | 259 | 154 | ||
BT002762.1 | - | 2 - 3 | 259 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
