Assay Details
Gene Symbol: ACL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085136.1 | NP_001078605.1 | 2 - 3 | 307 | 116 | |
NM_121958.4 | NP_568376.1 | 1 - 2 | 367 | 116 | ||
GenBank mRNA | AF184094.1 | - | 1 - 2 | 173 | 116 | |
AY087927.1 | - | 1 - 2 | 237 | 116 | ||
AY127003.1 | - | 1 - 2 | 242 | 116 | ||
BT002254.1 | - | 1 - 2 | 153 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
