Assay Details
Gene Symbol: ACL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085136.1 | NP_001078605.1 | 4 - 5 | 475 | 62 | |
NM_121958.4 | NP_568376.1 | 3 - 4 | 535 | 62 | ||
GenBank mRNA | AF184094.1 | - | 3 - 4 | 341 | 62 | |
AY087927.1 | - | 3 - 4 | 405 | 62 | ||
AY127003.1 | - | 3 - 4 | 410 | 62 | ||
BT002254.1 | - | 3 - 4 | 321 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
