Assay Details
Gene Symbol: ACL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085136.1 | NP_001078605.1 | 5 - 6 | 571 | 81 | |
NM_121958.4 | NP_568376.1 | 4 - 5 | 631 | 81 | ||
GenBank mRNA | AF184094.1 | - | 4 - 5 | 437 | 81 | |
AY087927.1 | - | 4 - 5 | 501 | 81 | ||
AY127003.1 | - | 4 - 5 | 506 | 81 | ||
BT002254.1 | - | 4 - 5 | 417 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
