Assay Details
Gene Symbol: ACL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085136.1 | NP_001078605.1 | 7 - 8 | 825 | 105 | |
NM_121958.4 | NP_568376.1 | 6 - 7 | 885 | 105 | ||
GenBank mRNA | AF184094.1 | - | 6 - 7 | 691 | 105 | |
AY087927.1 | - | 6 - 7 | 755 | 105 | ||
AY127003.1 | - | 6 - 7 | 760 | 105 | ||
BT002254.1 | - | 6 - 7 | 671 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
