Assay Details
Gene Symbol: DRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336273.1 | NP_001323803.1 | 4 - 5 | 954 | 85 | |
NM_001336274.1 | NP_001323802.1 | 3 - 4 | 1049 | 85 | ||
NM_128610.3 | NP_565702.1 | 3 - 4 | 1082 | 85 | ||
GenBank mRNA | AY045618.1 | - | 3 - 4 | 993 | 85 | |
BT000505.1 | - | 3 - 4 | 549 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
