Assay Details
Gene Symbol: GPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128714.4 | NP_180715.1 | 3 - 4 | 332 | 135 | |
GenBank mRNA | AY044330.1 | - | 3 - 4 | 184 | 135 | |
AY058187.1 | - | 3 - 4 | 287 | 135 | ||
AY086518.1 | - | 3 - 4 | 271 | 135 | ||
AY098982.1 | - | 3 - 4 | 184 | 135 | ||
U94495.1 | - | 3 - 4 | 209 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
