Assay Details
Gene Symbol: AT2G31790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128737.4 | NP_180738.1 | 1 - 2 | 359 | 127 | |
GenBank mRNA | AY056277.1 | - | 1 - 2 | 347 | 127 | |
AY117218.1 | - | 1 - 2 | 289 | 127 | ||
KJ138884.1 | - | 1 - 2 | 289 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
