Assay Details
Gene Symbol: BAM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128788.3 | NP_180788.2 | 4 - 5 | 1181 | 118 | |
GenBank mRNA | AY136463.1 | - | 4 - 5 | 1108 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
