Assay Details
Gene Symbol: KAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202728.1 | NP_001189657.1 | 3 - 4 | 954 | 75 | |
NM_128803.4 | NP_180803.1 | 2 - 3 | 812 | 75 | ||
GenBank mRNA | BT029168.1 | - | 2 - 3 | 714 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
