Assay Details
Gene Symbol: KAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202728.1 | NP_001189657.1 | 6 - 7 | 1384 | 65 | |
NM_128803.4 | NP_180803.1 | 5 - 6 | 1242 | 65 | ||
GenBank mRNA | BT029168.1 | - | 5 - 6 | 1144 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
