Assay Details
Gene Symbol: NUDT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336477.1 | NP_001325145.1 | 2 - 3 | 604 | 98 | |
NM_001336478.1 | NP_001325146.1 | 2 - 3 | 674 | 98 | ||
NM_001336479.1 | NP_001325147.1 | 2 - 3 | 584 | 98 | ||
NM_128949.3 | NP_565776.1 | 2 - 3 | 517 | 98 | ||
GenBank mRNA | AF360290.1 | - | 2 - 3 | 589 | 98 | |
AK175540.1 | - | 2 - 3 | 515 | 98 | ||
AK229595.1 | - | 2 - 3 | 551 | 98 | ||
AY051046.1 | - | 2 - 3 | 456 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
