Assay Details
Gene Symbol: AT2G34060 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128957.4 | NP_180953.1 | 1 - 2 | 373 | 98 | |
GenBank mRNA | BT002341.1 | - | 1 - 2 | 256 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
