Assay Details
Gene Symbol: AT2G34060 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128957.4 | NP_180953.1 | 2 - 3 | 583 | 78 | |
GenBank mRNA | BT002341.1 | - | 2 - 3 | 466 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
