Assay Details
Gene Symbol: AT1G51650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104043.3 | NP_175576.1 | 1 - 2 | 355 | 68 | |
GenBank mRNA | AY072309.1 | - | 1 - 2 | 248 | 68 | |
AY114540.1 | - | 1 - 2 | 183 | 68 | ||
D88377.1 | - | 1 - 2 | 226 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
