Assay Details
Gene Symbol: ROC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161212.1 | NP_001154684.1 | 4 - 5 | 401 | 75 | |
NM_116068.5 | NP_191762.1 | 2 - 3 | 384 | 75 | ||
GenBank mRNA | AY059843.1 | - | 2 - 3 | 305 | 75 | |
AY093284.1 | - | 2 - 3 | 242 | 75 | ||
L14845.1 | - | 2 - 3 | 269 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
