Assay Details
Gene Symbol: ROC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161212.1 | NP_001154684.1 | 5 - 6 | 559 | 67 | |
NM_001203230.1 | NP_001190159.1 | 3 - 4 | 541 | 67 | ||
NM_116068.5 | NP_191762.1 | 3 - 4 | 542 | 67 | ||
GenBank mRNA | AY059843.1 | - | 3 - 4 | 463 | 67 | |
AY093284.1 | - | 3 - 4 | 400 | 67 | ||
L14845.1 | - | 3 - 4 | 427 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
