Assay Details
Gene Symbol: ROC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161212.1 | NP_001154684.1 | 6 - 7 | 650 | 74 | |
NM_001203230.1 | NP_001190159.1 | 4 - 5 | 632 | 74 | ||
NM_116068.5 | NP_191762.1 | 4 - 5 | 633 | 74 | ||
GenBank mRNA | AY059843.1 | - | 4 - 5 | 554 | 74 | |
AY093284.1 | - | 4 - 5 | 491 | 74 | ||
L14845.1 | - | 4 - 5 | 518 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
