Assay Details
Gene Symbol: AT4G01890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340345.1 | NP_001319841.1 | 3 - 4 | 615 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
