Assay Details
Gene Symbol: MSH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203727.2 | NP_001190656.1 | 1 - 2 | 1156 | 125 | |
NM_116438.2 | NP_192116.1 | 1 - 2 | 1157 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
