Assay Details
Gene Symbol: SRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340445.1 | NP_001329389.1 | 12 - 13 | 2452 | 107 | |
NM_116577.5 | NP_192248.2 | 13 - 14 | 2374 | 107 | ||
GenBank mRNA | AY518288.1 | - | 13 - 14 | 2047 | 107 | |
FJ708743.1 | - | 12 - 13 | 1791 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
