Assay Details
Gene Symbol: SRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340445.1 | NP_001329389.1 | 8 - 9 | 1314 | 93 | |
NM_116577.5 | NP_192248.2 | 9 - 10 | 1236 | 93 | ||
GenBank mRNA | AY518288.1 | - | 9 - 10 | 909 | 93 | |
FJ708743.1 | - | 8 - 9 | 653 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
