Assay Details
Gene Symbol: EDA35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084885.2 | NP_001078354.1 | 1 | 581 | 65 | |
NM_116783.4 | NP_192453.1 | 1 | 615 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
