Assay Details
Gene Symbol: EHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116790.3 | NP_567299.2 | 10 - 11 | 835 | 66 | |
NM_202788.1 | NP_974517.1 | 10 - 11 | 784 | 66 | ||
GenBank mRNA | BT033036.1 | - | 10 - 11 | 716 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
