Assay Details
Gene Symbol: LysoPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104154.4 | NP_175685.1 | 2 - 3 | 892 | 121 | |
GenBank mRNA | AY054577.1 | - | 2 - 3 | 720 | 121 | |
BT008729.1 | - | 2 - 3 | 686 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
