Assay Details
Gene Symbol: AT1G54320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104310.4 | NP_564656.1 | 2 - 3 | 334 | 102 | |
GenBank mRNA | AF446869.1 | - | 2 - 3 | 146 | 102 | |
AY045672.1 | - | 2 - 3 | 310 | 102 | ||
AY053406.1 | - | 2 - 3 | 287 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
