Assay Details
Gene Symbol: AT1G54320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104310.4 | NP_564656.1 | 7 - 8 | 1186 | 65 | |
GenBank mRNA | AF446869.1 | - | 7 - 8 | 998 | 65 | |
AY045672.1 | - | 7 - 8 | 1162 | 65 | ||
AY053406.1 | - | 7 - 8 | 1139 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
