Assay Details
Gene Symbol: AT1G55290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104404.4 | NP_175925.1 | 1 - 2 | 627 | 98 | |
GenBank mRNA | BT011228.1 | - | 1 - 2 | 602 | 98 | |
BT012156.1 | - | 1 - 2 | 511 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
