Assay Details
Gene Symbol: TAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036117.1 | NP_001031194.1 | 3 - 4 | 572 | 103 | |
NM_104405.4 | NP_175926.1 | 3 - 4 | 572 | 103 | ||
GenBank mRNA | AK316930.1 | - | 3 - 4 | 553 | 103 | |
AK316996.1 | - | 3 - 4 | 534 | 103 | ||
AY463622.1 | - | 3 - 4 | 370 | 103 | ||
BT011002.1 | - | 3 - 4 | 497 | 103 | ||
BT015099.1 | - | 3 - 4 | 370 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
