Assay Details
Gene Symbol: TAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104405.4 | NP_175926.1 | 4 - 5 | 642 | 84 | |
GenBank mRNA | AK316930.1 | - | 4 - 5 | 623 | 84 | |
AY463622.1 | - | 4 - 5 | 440 | 84 | ||
BT011002.1 | - | 4 - 5 | 567 | 84 | ||
BT015099.1 | - | 4 - 5 | 440 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
