Assay Details
Gene Symbol: AAE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084254.1 | NP_001077723.1 | 10 - 11 | 1645 | 89 | |
NM_104408.5 | NP_175929.3 | 10 - 11 | 1908 | 89 | ||
GenBank mRNA | AY250845.1 | - | 10 - 11 | 1633 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
