Assay Details
Gene Symbol: AAE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084254.1 | NP_001077723.1 | 12 - 13 | 1861 | 104 | |
NM_104408.5 | NP_175929.3 | 12 - 13 | 2124 | 104 | ||
GenBank mRNA | AY250845.1 | - | 12 - 13 | 1849 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
