Assay Details
Gene Symbol: AAE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084254.1 | NP_001077723.1 | 2 - 3 | 407 | 137 | |
NM_104408.5 | NP_175929.3 | 2 - 3 | 674 | 137 | ||
GenBank mRNA | AY250845.1 | - | 2 - 3 | 399 | 137 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001084254.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
