Assay Details
Gene Symbol: AAE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084254.1 | NP_001077723.1 | 7 - 8 | 1113 | 111 | |
NM_104408.5 | NP_175929.3 | 7 - 8 | 1376 | 111 | ||
GenBank mRNA | AY250845.1 | - | 7 - 8 | 1101 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
