Assay Details
Gene Symbol: XF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104624.4 | NP_564734.1 | 2 - 3 | 495 | 66 | |
GenBank mRNA | AB008021.1 | - | 2 - 3 | 389 | 66 | |
AY099643.1 | - | 2 - 3 | 405 | 66 | ||
BT000239.1 | - | 2 - 3 | 361 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
