Assay Details
Gene Symbol: XF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104624.4 | NP_564734.1 | 5 - 6 | 1054 | 102 | |
GenBank mRNA | AB008021.1 | - | 5 - 6 | 948 | 102 | |
AY099643.1 | - | 5 - 6 | 964 | 102 | ||
BT000239.1 | - | 5 - 6 | 920 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
