Assay Details
Gene Symbol: EDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333862.1 | NP_001323259.1 | 2 - 3 | 511 | 59 | |
NM_104661.1 | NP_176177.1 | 2 - 3 | 352 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
