Assay Details
Gene Symbol: ARF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036131.3 | NP_001031208.1 | 14 - 15 | 1937 | 117 | |
NM_001198337.1 | NP_001185266.1 | 14 - 15 | 2168 | 117 | ||
NM_104668.4 | NP_176184.1 | 14 - 15 | 2133 | 117 | ||
NM_179499.2 | NP_849830.1 | 14 - 15 | 2079 | 117 | ||
GenBank mRNA | AK317214.1 | - | 14 - 15 | 1893 | 117 | |
BT002748.1 | - | 13 - 14 | 1822 | 117 | ||
U83245.1 | - | 14 - 15 | 1936 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
