Assay Details
Gene Symbol: ARL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333872.1 | NP_001320772.1 | 2 - 3 | 646 | 80 | |
NM_001333873.1 | NP_001320771.1 | 1 - 2 | 695 | 80 | ||
NM_104690.4 | NP_176206.2 | 2 - 3 | 646 | 80 | ||
GenBank mRNA | AK118901.1 | - | 2 - 3 | 542 | 80 | |
BT005502.1 | - | 2 - 3 | 266 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
