Assay Details
Gene Symbol: ARPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333892.1 | NP_001319276.1 | 5 - 6 | 862 | 67 | |
NM_104730.3 | NP_564757.1 | 5 - 6 | 849 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
