Assay Details
Gene Symbol: AT1G60770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104760.2 | NP_176276.1 | 1 - 2 | 290 | 86 | |
GenBank mRNA | BT002876.1 | - | 1 - 2 | 290 | 86 | |
BT004401.1 | - | 1 - 2 | 224 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
