Assay Details
Gene Symbol: NOGC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334040.1 | NP_001321124.1 | 1 - 2 | 587 | 95 | |
NM_104936.2 | NP_176446.1 | 1 - 2 | 634 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
