Assay Details
Gene Symbol: GPX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105025.4 | NP_564813.1 | 3 - 4 | 356 | 119 | |
GenBank mRNA | AK227611.1 | - | 3 - 4 | 286 | 119 | |
AY086991.1 | - | 3 - 4 | 200 | 119 | ||
BT003059.1 | - | 3 - 4 | 184 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
