Assay Details
Gene Symbol: AtMYB103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105065.3 | NP_176575.1 | 1 - 2 | 281 | 107 | |
GenBank mRNA | AF214116.2 | - | 1 - 2 | 132 | 107 | |
AK226651.1 | - | 1 - 2 | 281 | 107 | ||
AY519564.1 | - | 1 - 2 | 132 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
