Assay Details
Gene Symbol: AtMYB103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105065.3 | NP_176575.1 | 2 - 3 | 413 | 72 | |
GenBank mRNA | AF214116.2 | - | 2 - 3 | 264 | 72 | |
AK226651.1 | - | 2 - 3 | 413 | 72 | ||
AY519564.1 | - | 2 - 3 | 264 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
