Assay Details
Gene Symbol: AT1G64850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105159.4 | NP_564841.1 | 2 - 3 | 311 | 97 | |
GenBank mRNA | AY070488.1 | - | 2 - 3 | 179 | 97 | |
AY101542.1 | - | 2 - 3 | 152 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
