Assay Details
Gene Symbol: AT1G65240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105197.3 | NP_176703.1 | 7 - 8 | 1302 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
