Assay Details
Gene Symbol: NOV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117449.4 | NP_193111.2 | 5 - 6 | 2080 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
