Assay Details
Gene Symbol: AT4G15240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117612.3 | NP_193259.2 | 1 - 2 | 1054 | 88 | |
GenBank mRNA | KJ138758.1 | - | 1 - 2 | 814 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
